Assay Details
Gene Symbol: AT1G11630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101036.3 | NP_172629.1 | 1 | 1430 | 109 | |
GenBank mRNA | AF385742.1 | - | 1 | 1372 | 109 | |
AY087752.1 | - | 1 | 1363 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
