Assay Details
Gene Symbol: AT1G13410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101211.4 | NP_172798.2 | 2 | 1663 | 75 | |
GenBank mRNA | AK226699.1 | - | 2 | 1642 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
