Assay Details
Gene Symbol: ECH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106264.3 | NP_177742.2 | 7 - 8 | 511 | 123 | |
GenBank mRNA | AY070763.1 | - | 7 - 8 | 487 | 123 | |
AY093712.1 | - | 7 - 8 | 448 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
