Assay Details
Gene Symbol: MYB63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106569.4 | NP_178039.1 | 1 - 2 | 389 | 108 | |
GenBank mRNA | AK175344.1 | - | 1 - 2 | 151 | 108 | |
AY519572.1 | - | 1 - 2 | 137 | 108 | ||
BT033125.1 | - | 1 - 2 | 137 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
