Assay Details
Gene Symbol: MYB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111757.4 | NP_187534.1 | 1 - 2 | 445 | 83 | |
GenBank mRNA | AY550295.1 | - | 1 - 2 | 331 | 83 | |
BT026429.1 | - | 1 - 2 | 331 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
