Assay Details
Gene Symbol: MES17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111924.3 | NP_187698.1 | 1 - 2 | 423 | 68 | |
GenBank mRNA | AY063992.1 | - | 1 - 2 | 319 | 68 | |
AY096692.1 | - | 1 - 2 | 259 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
