Assay Details
Gene Symbol: AT3G28200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113739.3 | NP_189460.1 | 1 | 971 | 154 | |
GenBank mRNA | AY150382.1 | - | 1 | 866 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
