Assay Details
Gene Symbol: AT3G28560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113776.3 | NP_189497.1 | 1 | 904 | 95 | |
GenBank mRNA | DQ132691.1 | - | 1 | 904 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
