Assay Details
Gene Symbol: MES11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113902.4 | NP_189622.1 | 2 - 3 | 999 | 141 | |
GenBank mRNA | BT003149.1 | - | 2 - 3 | 624 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
