Assay Details
Gene Symbol: AT3G49190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114778.4 | NP_190488.1 | 2 - 3 | 823 | 95 | |
GenBank mRNA | AK117581.1 | - | 2 - 3 | 823 | 95 | |
BT005436.1 | - | 2 - 3 | 808 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
