Assay Details
Gene Symbol: MES12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117058.4 | NP_192728.2 | 4 - 5 | 1074 | 96 | |
GenBank mRNA | AK221404.1 | - | 4 - 5 | 915 | 96 | |
AY054619.1 | - | 4 - 5 | 986 | 96 | ||
AY081515.1 | - | 4 - 5 | 777 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
