Assay Details
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117086.4 | NP_192756.2 | 5 - 6 | 1184 | 83 | |
GenBank mRNA | BT015348.1 | - | 5 - 6 | 1087 | 83 | |
BT020454.1 | - | 5 - 6 | 1052 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
