Assay Details
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117086.4 | NP_192756.2 | 6 - 7 | 1272 | 111 | |
GenBank mRNA | BT015348.1 | - | 6 - 7 | 1175 | 111 | |
BT020454.1 | - | 6 - 7 | 1140 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
