Assay Details
Gene Symbol: EIF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117213.4 | NP_192881.1 | 12 - 13 | 2407 | 70 | |
GenBank mRNA | AF291711.1 | - | 12 - 13 | 2322 | 70 | |
AK226553.1 | - | 12 - 13 | 2365 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
