Assay Details
Gene Symbol: EIF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117213.4 | NP_192881.1 | 13 - 14 | 2612 | 69 | |
GenBank mRNA | AF291711.1 | - | 13 - 14 | 2527 | 69 | |
AK226553.1 | - | 13 - 14 | 2570 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
