Assay Details
Gene Symbol: MYB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341904.1 | NP_001328222.1 | 1 - 2 | 264 | 82 | |
NM_118951.3 | NP_194540.1 | 1 - 2 | 273 | 82 | ||
GenBank mRNA | AY519610.1 | - | 1 - 2 | 132 | 82 | |
BT001930.1 | - | 1 - 2 | 132 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001341904.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
