Assay Details
Gene Symbol: AT5G25590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122470.3 | NP_197941.1 | 1 - 2 | 1327 | 132 | |
GenBank mRNA | AY091053.1 | - | 1 - 2 | 1184 | 132 | |
AY133786.1 | - | 1 - 2 | 1111 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
