Assay Details
Gene Symbol: AT5G26810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343983.1 | NP_001318658.1 | 3 - 4 | 708 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
