Assay Details
Gene Symbol: MYB116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102344.3 | NP_564230.1 | 1 - 2 | 338 | 105 | |
GenBank mRNA | AF334815.2 | - | 1 - 2 | 277 | 105 | |
AY519558.1 | - | 1 - 2 | 148 | 105 | ||
DQ446290.1 | - | 1 - 2 | 148 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
