Assay Details
Gene Symbol: AT1G73560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106014.2 | NP_565067.1 | 1 - 2 | 343 | 69 | |
GenBank mRNA | DQ446425.1 | - | 1 - 2 | 328 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
