Assay Details
Gene Symbol: AT1G73740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106033.4 | NP_177515.1 | 3 - 4 | 1603 | 65 | |
GenBank mRNA | AY059826.1 | - | 3 - 4 | 1348 | 65 | |
AY064640.1 | - | 3 - 4 | 1176 | 65 | ||
KJ138939.1 | - | 3 - 4 | 1176 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
