Assay Details
Gene Symbol: MYB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123397.3 | NP_198849.1 | 2 - 3 | 383 | 64 | |
GenBank mRNA | AY519631.1 | - | 2 - 3 | 259 | 64 | |
BT025285.1 | - | 2 - 3 | 259 | 64 | ||
Z68158.1 | - | 2 - 3 | 379 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
