Assay Details
Gene Symbol: AT5G41800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123545.4 | NP_568597.1 | 6 - 7 | 1146 | 99 | |
GenBank mRNA | AY099685.1 | - | 6 - 7 | 978 | 99 | |
AY128864.1 | - | 6 - 7 | 967 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
