Assay Details
Gene Symbol: NRPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344590.1 | NP_001332060.1 | 35 - 36 | 3374 | 82 | |
NM_123882.5 | NP_199327.4 | 35 - 36 | 3292 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
