Assay Details
Gene Symbol: AT5G47390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124110.4 | NP_199550.1 | 2 - 3 | 717 | 89 | |
GenBank mRNA | AK226931.1 | - | 2 - 3 | 635 | 89 | |
AY072077.1 | - | 2 - 3 | 642 | 89 | ||
AY084295.1 | - | 2 - 3 | 597 | 89 | ||
AY096661.1 | - | 2 - 3 | 485 | 89 | ||
AY519516.1 | - | 2 - 3 | 485 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
