Assay Details
Gene Symbol: PME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124123.5 | NP_199561.1 | 2 - 3 | 530 | 80 | |
GenBank mRNA | AY102117.1 | - | 2 - 3 | 512 | 80 | |
BT000565.1 | - | 2 - 3 | 483 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
