Assay Details
Gene Symbol: PME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124123.5 | NP_199561.1 | 3 - 4 | 744 | 114 | |
GenBank mRNA | AY102117.1 | - | 3 - 4 | 726 | 114 | |
BT000565.1 | - | 3 - 4 | 697 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
