Assay Details
Gene Symbol: PME5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124123.5 | NP_199561.1 | 4 - 5 | 982 | 68 | |
GenBank mRNA | AY102117.1 | - | 4 - 5 | 964 | 68 | |
BT000565.1 | - | 4 - 5 | 935 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
