Assay Details
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161296.1 | NP_001154768.1 | 16 - 17 | 2334 | 127 | |
NM_001344757.1 | NP_001318757.1 | 17 - 18 | 2375 | 127 | ||
NM_124183.4 | NP_199620.4 | 16 - 17 | 2200 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
