Assay Details
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161296.1 | NP_001154768.1 | 17 - 18 | 2392 | 129 | |
NM_001344757.1 | NP_001318757.1 | 18 - 19 | 2433 | 129 | ||
NM_124183.4 | NP_199620.4 | 17 - 18 | 2258 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
