Assay Details
Gene Symbol: CPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203575.1 | NP_001190504.1 | 5 - 6 | 565 | 80 | |
NM_001344891.1 | NP_001331972.1 | 4 - 5 | 840 | 80 | ||
NM_124419.5 | NP_568727.1 | 5 - 6 | 590 | 80 | ||
GenBank mRNA | AF216756.1 | - | 5 - 6 | 408 | 80 | |
BT025804.1 | - | 4 - 5 | 376 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
