Assay Details
Gene Symbol: FOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124718.4 | NP_200151.2 | 1 - 2 | 357 | 61 | |
GenBank mRNA | AB189175.1 | - | 1 - 2 | 246 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
