Assay Details
Gene Symbol: PLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124738.4 | NP_200170.2 | 2 - 3 | 436 | 112 | |
GenBank mRNA | AK221481.1 | - | 2 - 3 | 402 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
