Assay Details
Gene Symbol: PLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124738.4 | NP_200170.2 | 4 - 5 | 652 | 89 | |
GenBank mRNA | AK221481.1 | - | 4 - 5 | 618 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
