Assay Details
Gene Symbol: SBE2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129196.4 | NP_181180.1 | 3 - 4 | 601 | 131 | |
GenBank mRNA | AK226896.1 | - | 3 - 4 | 552 | 131 | |
AY136411.1 | - | 3 - 4 | 575 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
