Assay Details
Gene Symbol: AKR4C10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129335.5 | NP_181315.2 | 2 - 3 | 323 | 109 | |
GenBank mRNA | AK227354.1 | - | 2 - 3 | 228 | 109 | |
BT005862.1 | - | 2 - 3 | 177 | 109 | ||
DQ837655.1 | - | 2 - 3 | 177 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
