Assay Details
Gene Symbol: AKR4C10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129335.5 | NP_181315.2 | 4 - 5 | 489 | 97 | |
GenBank mRNA | DQ837655.1 | - | 4 - 5 | 343 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
