Assay Details
Gene Symbol: AKR4C10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129335.5 | NP_181315.2 | 6 - 7 | 1001 | 68 | |
GenBank mRNA | AK227354.1 | - | 6 - 7 | 906 | 68 | |
BT005862.1 | - | 6 - 7 | 855 | 68 | ||
DQ837655.1 | - | 6 - 7 | 855 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
