Assay Details
Gene Symbol: PXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129555.4 | NP_181526.1 | 4 - 5 | 561 | 71 | |
GenBank mRNA | AB047148.1 | - | 3 - 4 | 230 | 71 | |
AY039858.1 | - | 4 - 5 | 378 | 71 | ||
BT003020.1 | - | 3 - 4 | 230 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
