Assay Details
Gene Symbol: PXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129555.4 | NP_181526.1 | 7 - 8 | 856 | 69 | |
GenBank mRNA | AB047148.1 | - | 6 - 7 | 525 | 69 | |
AY039858.1 | - | 7 - 8 | 673 | 69 | ||
BT003020.1 | - | 6 - 7 | 525 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
