Assay Details
Gene Symbol: PXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129555.4 | NP_181526.1 | 9 - 10 | 1025 | 58 | |
GenBank mRNA | AB047148.1 | - | 8 - 9 | 694 | 58 | |
AY039858.1 | - | 9 - 10 | 842 | 58 | ||
BT003020.1 | - | 8 - 9 | 694 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
