Assay Details
Gene Symbol: AT2G40250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129583.2 | NP_181554.1 | 2 - 3 | 932 | 117 | |
GenBank mRNA | DQ056573.1 | - | 2 - 3 | 899 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
