Assay Details
Gene Symbol: AT2G41090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129674.4 | NP_181642.1 | 1 - 2 | 553 | 63 | |
GenBank mRNA | AY046044.1 | - | 1 - 2 | 116 | 63 | |
AY084987.1 | - | 1 - 2 | 116 | 63 | ||
Z12136.1 | - | 91 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
