Assay Details
Gene Symbol: AT2G41090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129674.4 | NP_181642.1 | 2 - 3 | 917 | 72 | |
GenBank mRNA | AY046044.1 | - | 2 - 3 | 480 | 72 | |
AY084987.1 | - | 2 - 3 | 480 | 72 | ||
Z12136.1 | - | 455 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
