Assay Details
Gene Symbol: AT2G42490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129810.5 | NP_181777.2 | 10 - 11 | 2611 | 81 | |
GenBank mRNA | AY120717.1 | - | 10 - 11 | 2436 | 81 | |
BT000029.1 | - | 10 - 11 | 2114 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
