Assay Details
Gene Symbol: AT2G42490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129810.5 | NP_181777.2 | 4 - 5 | 1226 | 86 | |
GenBank mRNA | AY120717.1 | - | 4 - 5 | 1051 | 86 | |
BT000029.1 | - | 4 - 5 | 729 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
