Assay Details
Gene Symbol: AT2G42490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129810.5 | NP_181777.2 | 9 - 10 | 2335 | 66 | |
GenBank mRNA | AY120717.1 | - | 9 - 10 | 2160 | 66 | |
BT000029.1 | - | 9 - 10 | 1838 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
