Assay Details
Gene Symbol: AT2G43500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161101.2 | NP_001154573.1 | 5 - 6 | 1960 | 95 | |
NM_001337037.1 | NP_001318414.1 | 5 - 6 | 2130 | 95 | ||
NM_001337038.1 | NP_001325296.1 | 3 - 4 | 2374 | 95 | ||
NM_001337039.1 | NP_001325295.1 | 4 - 5 | 2280 | 95 | ||
NM_001337040.1 | NP_001325294.1 | 4 - 5 | 2224 | 95 | ||
NM_001337041.1 | NP_001325293.1 | 6 - 7 | 2080 | 95 | ||
NM_001337042.1 | NP_001325292.1 | 6 - 7 | 2076 | 95 | ||
NM_001337043.1 | NP_001325291.1 | 5 - 6 | 2037 | 95 | ||
NM_001337044.1 | NP_001325290.1 | 5 - 6 | 2005 | 95 | ||
NM_001337045.1 | NP_001325289.1 | 5 - 6 | 1959 | 95 | ||
NM_001337046.1 | NP_001325288.1 | 5 - 6 | 2031 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
