Assay Details
Gene Symbol: MES19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335855.1 | NP_001318275.1 | 2 - 3 | 526 | 150 | |
GenBank mRNA | DQ056539.1 | - | 2 - 3 | 262 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
