Assay Details
Gene Symbol: ACL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335856.1 | NP_001323663.1 | 1 - 2 | 577 | 165 | |
NM_001335857.1 | NP_001323664.1 | 1 - 2 | 500 | 165 | ||
NM_127924.3 | NP_179941.1 | 1 - 2 | 410 | 165 | ||
GenBank mRNA | AF361627.1 | - | 1 - 2 | 391 | 165 | |
AK226527.1 | - | 1 - 2 | 396 | 165 | ||
AY056068.1 | - | 1 - 2 | 378 | 165 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
